missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Ly-6A/E Antibody (SR3456), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals™ NBP3-27773-50ul
This item is not returnable.
View return policy
Description
Ly-6A/E Monoclonal antibody specifically detects Ly-6A/E in Mouse samples. It is validated for Immunohistochemistry,Western Blot
Specifications
| Ly-6A/E | |
| Monoclonal | |
| Unconjugated | |
| PBS (pH 7.4), 150mM NaCl, 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Mouse | |
| Purified |
| Immunohistochemistry, Western Blot | |
| SR3456 | |
| Western Blot 1:500-1:2000, Immunohistochemistry 1:50-1:200 | |
| Ly 6A/E, Ly6A.2, Ly6A/E, Ly6E.1, Lymphocyte antigen 6 complex locus A, Lymphocyte antigen 6 complex locus E, Retinoic acid induced gene E, RIGE, SCA1, SCA-1, SCA2, Stem cell antigen 1, Stem cell antigen 2, T cell activating protein, TAP, Thymic shared antigen 1, TSA1 | |
| A synthesized peptide derived from human Ly-6A/E (Uniprot #: P05533) | |
| 50 μL | |
| Hematopoietic Stem Cell Markers, Stem Cell Markers | |
| 110454 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction