missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Ly-6A/E Antibody (SR3456), Novus Biologicals™
Rabbit Monoclonal Antibody
327.00€ - 382.00€
Specifications
| Antigen | Ly-6A/E |
|---|---|
| Clone | SR3456 |
| Dilution | Western Blot 1:500-1:2000, Immunohistochemistry 1:50-1:200 |
| Applications | Immunohistochemistry, Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30041524
|
Novus Biologicals™
NBP3-27773-50ul |
50 μL |
327.00€
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30041602
|
Novus Biologicals™
NBP3-27773-100ul |
100 μL |
382.00€
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Ly-6A/E Monoclonal antibody specifically detects Ly-6A/E in Mouse samples. It is validated for Immunohistochemistry,Western BlotSpecifications
| Ly-6A/E | |
| Western Blot 1:500-1:2000, Immunohistochemistry 1:50-1:200 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Mouse | |
| Ly 6A/E, Ly6A.2, Ly6A/E, Ly6E.1, Lymphocyte antigen 6 complex locus A, Lymphocyte antigen 6 complex locus E, Retinoic acid induced gene E, RIGE, SCA1, SCA-1, SCA2, Stem cell antigen 1, Stem cell antigen 2, T cell activating protein, TAP, Thymic shared antigen 1, TSA1 | |
| A synthesized peptide derived from human Ly-6A/E (Uniprot #: P05533) | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. |
| SR3456 | |
| Immunohistochemistry, Western Blot | |
| Unconjugated | |
| Rabbit | |
| Hematopoietic Stem Cell Markers, Stem Cell Markers | |
| PBS (pH 7.4), 150mM NaCl, 50% glycerol | |
| 110454 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title