Learn More
Lutetium(III) bromide, ultra dry, 99.99% (metals basis), Thermo Scientific™
80.40€ - 267.00€
Chemical Identifiers
CAS 14456-53-2
MDL Number MFCD00049965
View More Specs
Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15407465
SDS View Documents
Thermo Scientific Alfa Aesar
047190.03
1g
80.40€
1g
View alternative products
Save Up to 
»
15417465
SDS View Documents
Thermo Scientific Alfa Aesar
047190.06
5g
267.00€
5g
View alternative products
Save Up to 
»
Description
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
  • Ampouled under argon
Specifications
Chemical Identifiers
14456-53-2
MFCD00049965
Specifications
14456-53-2
414.68
Odorless
Hygroscopic
Lutetium(III) bromide, Ultra dry
MFCD00049965
99.99%
Ampule
(metals basis)
Videos
Safety and Handling
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
missing translation for 'einecsNumber' : 238-446-5
missing translation for 'tsca' : Yes
missing translation for 'storageNote' :
  • Ampouled under argon
Recommended Storage : Ambient temperatures; Store under Nitrogen