accessibility menu, dialog, popup

UserName

CAS RN 14456-53-2

BrBrBrLu

CAS RN 14456-53-2

IUPAC Name: lutetium(3+) tribromide
Synonyms: lutetium(3+) tribromide
Molecular Weight (g/mol): 414.68
Molecular Formula: Br3Lu
InChi Key: DWHGOINJUKABSY-UHFFFAOYSA-K
SMILES: [Br-].[Br-].[Br-].[Lu+3]
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)