Learn More
Lutetium(III) bromide, ultra dry, 99.99% (metals basis), Thermo Scientific™
Brand: Thermo Scientific Alfa Aesar 047190.06
| Quantity | 5g |
|---|
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
- Ampouled under argon
Chemical Identifiers
| 14456-53-2 | |
| MFCD00049965 |
Specifications
| 14456-53-2 | |
| MFCD00049965 | |
| 99.99% | |
| Ampule | |
| (metals basis) |
| 5g | |
| 414.68 | |
| Odorless | |
| Hygroscopic | |
| Lutetium(III) bromide, Ultra dry |