missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LRRC32/GARP Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 2 publications
236.00€ - 372.00€
Specifications
| Antigen | LRRC32/GARP |
|---|---|
| Concentration | 1.0 mg/mL |
| Dilution | Western Blot 0.5-1.0 ug/ml, Flow Cytometry 1-3 ug/ml |
| Applications | Western Blot, Flow Cytometry |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18094304
|
Novus Biologicals
NBP2-24664SS |
0.025 mg |
236.00€
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18776103
|
Novus Biologicals
NBP2-24664 |
0.1 mg |
372.00€
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
LRRC32/GARP Polyclonal specifically detects LRRC32/GARP in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry.Specifications
| LRRC32/GARP | |
| Western Blot 0.5-1.0 ug/ml, Flow Cytometry 1-3 ug/ml | |
| Polyclonal | |
| Purified | |
| RUO | |
| 0.1 ml PBS with 0.05% Sodium Azide | |
| D11S833Eleucine-rich repeat-containing protein 32, GARPGarpin, Glycoprotein A repetitions predominantgarpin, leucine rich repeat containing 32 | |
| LRRC32 | |
| IgG | |
| Protein A purified | |
| The amino acid sequence used as immunogen is 100% homologous in human (isoform CRA_a), chicken, cow, dog, mouse, rat and monkey, and 92% homologous in xenopus. FOXO3A is a 673 amino acid protein which is observed at approximately 95 kD in western blot. |
| 1.0 mg/mL | |
| Western Blot, Flow Cytometry | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse, Rat | |
| Q14392 | |
| 2615 | |
| A portion of amino acids 250-300 of human GARP/LRRC32 was used as the immunogen. | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title