missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LRRC32/GARP Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 2 publications
Brand: Novus Biologicals NBP2-24664
This item is not returnable.
View return policy
Description
LRRC32/GARP Polyclonal specifically detects LRRC32/GARP in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry.
Specifications
| LRRC32/GARP | |
| Polyclonal | |
| Unconjugated | |
| 0.1 ml PBS with 0.05% Sodium Azide | |
| D11S833Eleucine-rich repeat-containing protein 32, GARPGarpin, Glycoprotein A repetitions predominantgarpin, leucine rich repeat containing 32 | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| 2615 | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, Flow Cytometry | |
| 1.0 mg/mL | |
| Western Blot 0.5-1.0 ug/ml, Flow Cytometry 1-3 ug/ml | |
| Q14392 | |
| LRRC32 | |
| A portion of amino acids 250-300 of human GARP/LRRC32 was used as the immunogen. | |
| 0.1 mg | |
| Primary | |
| The amino acid sequence used as immunogen is 100% homologous in human (isoform CRA_a), chicken, cow, dog, mouse, rat and monkey, and 92% homologous in xenopus. FOXO3A is a 673 amino acid protein which is observed at approximately 95 kD in western blot. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction