missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LRP-4 Antibody (S207-27), Novus Biologicals™
Mouse Monoclonal Antibody
178.00€
Specifications
| Antigen | LRP-4 |
|---|---|
| Clone | S207-27 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1μg/mL, Immunocytochemistry/Immunofluorescence 1:10-1:2000 |
| Applications | Western Blot, Immunocytochemistry, Immunofluorescence |
Description
LRP-4 Monoclonal specifically detects LRP-4 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.Specifications
| LRP-4 | |
| 1 mg/mL | |
| Western Blot, Immunocytochemistry, Immunofluorescence | |
| Unconjugated | |
| Mouse | |
| Human, Mouse, Rat | |
| CLSS, KIAA0816, low density lipoprotein receptor-related protein 4, low-density lipoprotein receptor-related protein 4, LRP-4, MEGF7LRP10, Multiple epidermal growth factor-like domains 7 | |
| LRP4 | |
| IgG2a | |
| Protein G purified | |
| Detects the extracellular domain of LRP4 approx 215kDa. Degradation product can be seen at approx 150kDa. |
| S207-27 | |
| Western Blot 1μg/mL, Immunocytochemistry/Immunofluorescence 1:10-1:2000 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Q8VI56 | |
| 4038 | |
| Fusion protein amino acids 26- 350 (extracellular N-terminus) of mouse LTP4 | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title