missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LRP-4 Antibody (S207-27), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-22416-0.025mg
This item is not returnable.
View return policy
Description
LRP-4 Monoclonal specifically detects LRP-4 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.
Specifications
| LRP-4 | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1μg/mL, Immunocytochemistry/Immunofluorescence 1:10-1:2000 | |
| CLSS, KIAA0816, low density lipoprotein receptor-related protein 4, low-density lipoprotein receptor-related protein 4, LRP-4, MEGF7LRP10, Multiple epidermal growth factor-like domains 7 | |
| Mouse | |
| Protein G purified | |
| RUO | |
| 4038 | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, Immunocytochemistry, Immunofluorescence | |
| S207-27 | |
| Unconjugated | |
| Q8VI56 | |
| LRP4 | |
| Fusion protein amino acids 26- 350 (extracellular N-terminus) of mouse LTP4 | |
| 0.025 mg | |
| Primary | |
| Detects the extracellular domain of LRP4 approx 215kDa. Degradation product can be seen at approx 150kDa. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction