missing translation for 'onlineSavingsMsg'
Learn More

LONRF3 Antibody (OTI7F7), Novus Biologicals Biologicals™

Mouse Monoclonal Antibody

256.00€ - 550.00€

Specifications

Antigen LONRF3
Clone OTI7F7
Dilution Western Blot 1:500
Applications Western Blot
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
30306358
View Documents
Novus Biologicals
NBP3-44961-30ul
30 μL
256.00€
30µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
30306627
View Documents
Novus Biologicals
NBP3-44961-100ul
100 μL
550.00€
100µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

LONRF3 Monoclonal antibody specifically detects LONRF3 in Human samples. It is validated for Western Blot
Specifications

Specifications

LONRF3
Western Blot 1:500
Monoclonal
Purified
RUO
Human
LON peptidase N-terminal domain and ring finger 3, LON peptidase N-terminal domain and RING finger protein 3, MGC119463, MGC119465, RING finger protein 127FLJ22612, RNF127
Human recombinant protein fragment corresponding to amino acids 206-456 of human LONRF3 (NP_001027026) produced in E.coli.
Primary
Store at -20 degrees C. Avoid freeze/thaw cycles.
OTI7F7
Western Blot
Unconjugated
Mouse
Zinc Finger
PBS (pH 7.3) 1% BSA, 50% glycerol
79836
IgG1
>95%
Antibody
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.