Learn More
LONRF3 Antibody (OTI7F7), Novus Biologicals Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-44961-100ul
This item is not returnable.
View return policy
Description
LONRF3 Monoclonal antibody specifically detects LONRF3 in Human samples. It is validated for Western Blot
Specifications
| LONRF3 | |
| Monoclonal | |
| Unconjugated | |
| PBS (pH 7.3) 1% BSA, 50% glycerol | |
| Mouse | |
| >95% | |
| RUO | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Western Blot | |
| OTI7F7 | |
| Western Blot 1:500 | |
| LON peptidase N-terminal domain and ring finger 3, LON peptidase N-terminal domain and RING finger protein 3, MGC119463, MGC119465, RING finger protein 127FLJ22612, RNF127 | |
| Human recombinant protein fragment corresponding to amino acids 206-456 of human LONRF3 (NP_001027026) produced in E.coli. | |
| 100 μL | |
| Zinc Finger | |
| 79836 | |
| Store at -20 degrees C. Avoid freeze/thaw cycles. | |
| Purified |