missing translation for 'onlineSavingsMsg'
Learn More

Lofexidine hydrochloride, 98+%, Thermo Scientific™

Alpha-2 adrenergic agonist

130.00€ - 499.00€

Chemical Identifiers

CAS 21498-08-8
Molecular Formula C11H13Cl3N2O
Molecular Weight (g/mol) 295.588
MDL Number MFCD00917022
InChI Key DWWHMKBNNNZGHF-UHFFFAOYSA-N
Synonym lofexidine hydrochloride, lofexidine hcl, lofetensin, loxacor, lofexidine.hcl, loxacor hydrochloride, lofetensin hydrochloride, 2-1-2,6-dichlorophenoxy ethyl-4,5-dihydro-1h-imidazole hydrochloride, lofexidine hydrochloride usan
PubChem CID 30667
IUPAC Name 2-[1-(2,6-dichlorophenoxy)ethyl]-4,5-dihydro-1H-imidazole;hydrochloride
SMILES CC(C1=NCCN1)OC2=C(C=CC=C2Cl)Cl.Cl
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15446109
SDS View Documents
Thermo Scientific Alfa Aesar
J63960.03
1g
130.00€
1g
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
15456109
SDS View Documents
Thermo Scientific Alfa Aesar
J63960.06
5g
499.00€
5g
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

21498-08-8
295.588
DWWHMKBNNNZGHF-UHFFFAOYSA-N
30667
CC(C1=NCCN1)OC2=C(C=CC=C2Cl)Cl.Cl
C11H13Cl3N2O
MFCD00917022
lofexidine hydrochloride, lofexidine hcl, lofetensin, loxacor, lofexidine.hcl, loxacor hydrochloride, lofetensin hydrochloride, 2-1-2,6-dichlorophenoxy ethyl-4,5-dihydro-1h-imidazole hydrochloride, lofexidine hydrochloride usan
2-[1-(2,6-dichlorophenoxy)ethyl]-4,5-dihydro-1H-imidazole;hydrochloride

Specifications

21498-08-8
MFCD00917022
14,5558
DWWHMKBNNNZGHF-UHFFFAOYSA-N
2-[1-(2,6-dichlorophenoxy)ethyl]-4,5-dihydro-1H-imidazole;hydrochloride
30667
≥98%
C11H13Cl3N2O
UN2811
lofexidine hydrochloride, lofexidine hcl, lofetensin, loxacor, lofexidine.hcl, loxacor hydrochloride, lofetensin hydrochloride, 2-1-2,6-dichlorophenoxy ethyl-4,5-dihydro-1h-imidazole hydrochloride, lofexidine hydrochloride usan
CC(C1=NCCN1)OC2=C(C=CC=C2Cl)Cl.Cl
295.588
295.6
Lofexidine hydrochloride
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H301
Toxic if swallowed.

P264b-P270-P301+P310-P330-P501c

H301

missing translation for 'dotInformation' : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.

missing translation for 'tsca' : No

Recommended Storage : Keep cold

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.