Learn More
Lofexidine hydrochloride, 98+%, Thermo Scientific™
Alpha-2 adrenergic agonist
Brand: Thermo Scientific Alfa Aesar J63960.03
| Quantity | 1g |
|---|
View more versions of this product
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
| 21498-08-8 | |
| 295.588 | |
| DWWHMKBNNNZGHF-UHFFFAOYSA-N | |
| 30667 | |
| CC(C1=NCCN1)OC2=C(C=CC=C2Cl)Cl.Cl |
| C11H13Cl3N2O | |
| MFCD00917022 | |
| lofexidine hydrochloride, lofexidine hcl, lofetensin, loxacor, lofexidine.hcl, loxacor hydrochloride, lofetensin hydrochloride, 2-1-2,6-dichlorophenoxy ethyl-4,5-dihydro-1h-imidazole hydrochloride, lofexidine hydrochloride usan | |
| 2-[1-(2,6-dichlorophenoxy)ethyl]-4,5-dihydro-1H-imidazole;hydrochloride |
Specifications
| 21498-08-8 | |
| MFCD00917022 | |
| UN2811 | |
| lofexidine hydrochloride, lofexidine hcl, lofetensin, loxacor, lofexidine.hcl, loxacor hydrochloride, lofetensin hydrochloride, 2-1-2,6-dichlorophenoxy ethyl-4,5-dihydro-1h-imidazole hydrochloride, lofexidine hydrochloride usan | |
| CC(C1=NCCN1)OC2=C(C=CC=C2Cl)Cl.Cl | |
| 295.588 | |
| 295.6 | |
| Lofexidine hydrochloride |
| C11H13Cl3N2O | |
| 1g | |
| 14,5558 | |
| DWWHMKBNNNZGHF-UHFFFAOYSA-N | |
| 2-[1-(2,6-dichlorophenoxy)ethyl]-4,5-dihydro-1H-imidazole;hydrochloride | |
| 30667 | |
| ≥98% |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.