accessibility menu, dialog, popup

UserName

HClCH3OClClNHN

CAS RN 21498-08-8

IUPAC Name: hydrogen 2-[1-(2,6-dichlorophenoxy)ethyl]-4,5-dihydro-1H-imidazole chloride
Synonyms: lofexidine
Molecular Weight (g/mol): 295.59
Molecular Formula: C11H13Cl3N2O
InChi Key: DWWHMKBNNNZGHF-UHFFFAOYNA-N
SMILES: [H+].[Cl-].CC(OC1=C(Cl)C=CC=C1Cl)C1=NCCN1
Molecular Weight (g/mol) 
  • (2)
  • (5)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Certifications/Compliance 
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (2)
  • (3)
  • (2)
  • (5)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)