missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Lnx1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
213.00€ - 508.00€
Specifications
| Antigen | Lnx1 |
|---|---|
| Dilution | Western Blot : 1:400 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30067108
|
Novus Biologicals™
NBP3-30915-100ug |
100 μg |
508.00€
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30066795
|
Novus Biologicals™
NBP3-30915-25ug |
25 μg |
213.00€
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Lnx1 Polyclonal antibody specifically detects Lnx1 in Human samples. It is validated for Western BlotSpecifications
| Lnx1 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Cancer, Hypoxia, Lipid and Metabolism, Zinc Finger | |
| Phosphate buffered saline | |
| 84708 | |
| Primary | |
| Store at -20°C. Avoid freeze/thaw cycles. |
| Western Blot : 1:400 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human | |
| EC 6.3.2.-, ligand of numb-protein X 1ligand of numb-protein X, LNXmulti-PDZ-domain-containing protein, E3 ubiquitin-protein ligase LNX, MPDZ, Numb-binding protein 1, PDZ domain-containing ring finger protein 2, PDZRN2E3 ubiquitin-protein ligase LNX | |
| Synthetic peptide derived from the human Lnx1 protein. | |
| Antigen Affinity-purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title