missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Lnx1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals™ NBP3-30915-100ug
This item is not returnable.
View return policy
Description
Lnx1 Polyclonal antibody specifically detects Lnx1 in Human samples. It is validated for Western Blot
Specifications
| Lnx1 | |
| Polyclonal | |
| Western Blot : 1:400 | |
| EC 6.3.2.-, ligand of numb-protein X 1ligand of numb-protein X, LNXmulti-PDZ-domain-containing protein, E3 ubiquitin-protein ligase LNX, MPDZ, Numb-binding protein 1, PDZ domain-containing ring finger protein 2, PDZRN2E3 ubiquitin-protein ligase LNX | |
| Synthetic peptide derived from the human Lnx1 protein. | |
| 100 μg | |
| Cancer, Hypoxia, Lipid and Metabolism, Zinc Finger | |
| 84708 | |
| Store at -20°C. Avoid freeze/thaw cycles. |
| Western Blot | |
| Unconjugated | |
| Phosphate buffered saline | |
| Rabbit | |
| Antigen Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction