missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ LMW-PTP/ACP1 Antibody, Novus Biologicals™
Sheep Polyclonal Antibody
213.00€ - 508.00€
Specifications
| Antigen | LMW-PTP/ACP1 |
|---|---|
| Dilution | Western Blot : 1-10 μg/mL, Immunoprecipitation : 1-10 μg/mL |
| Applications | Immunoprecipitation, Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30066971
|
Novus Biologicals™
NBP3-30788-25ug |
25 μg |
213.00€
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30066785
|
Novus Biologicals™
NBP3-30788-100ug |
100 μg |
508.00€
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
LMW-PTP/ACP1 Polyclonal antibody specifically detects LMW-PTP/ACP1 in Bovine,Human,Mouse,Rat samples. It is validated for Immunoprecipitation,Western BlotSpecifications
| LMW-PTP/ACP1 | |
| Immunoprecipitation, Western Blot | |
| Unconjugated | |
| Sheep | |
| Protein Phosphatase | |
| Phosphate buffered saline | |
| 52 | |
| IgG | |
| Ammonium sulfate precipitation |
| Western Blot : 1-10 μg/mL, Immunoprecipitation : 1-10 μg/mL | |
| Polyclonal | |
| Purified | |
| RUO | |
| Bovine, Human, Mouse, Rat | |
| acid phosphatase 1, soluble, acid phosphatase of erythrocyte, Adipocyte acid phosphatase, cytoplasmic phosphotyrosyl protein phosphatase, EC 3.1.3.2, EC 3.1.3.48, HAAP, LMW-PTP, LMW-PTPase, Low molecular weight cytosolic acid phosphatase, low molecular weight phosphotyrosine protein phosphatase, MGC111030, MGC3499, protein tyrosine phosphatase, Red cell acid phosphatase 1 | |
| The human protein, LMW-PTP/ACP1 (GB Accession #M83653) was subcloned into pRSET to express a His6 N terminal fusion protein. This protein was expressed in E. coli and purified to homogeneity on Ni affinity matrix. | |
| Primary | |
| Store at -20°C. Avoid freeze/thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title