missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ LMW-PTP/ACP1 Antibody, Novus Biologicals™
Sheep Polyclonal Antibody
Brand: Novus Biologicals™ NBP3-30788-25ug
This item is not returnable.
View return policy
Description
LMW-PTP/ACP1 Polyclonal antibody specifically detects LMW-PTP/ACP1 in Bovine,Human,Mouse,Rat samples. It is validated for Immunoprecipitation,Western Blot
Specifications
| LMW-PTP/ACP1 | |
| Polyclonal | |
| Western Blot : 1-10 μg/mL, Immunoprecipitation : 1-10 μg/mL | |
| acid phosphatase 1, soluble, acid phosphatase of erythrocyte, Adipocyte acid phosphatase, cytoplasmic phosphotyrosyl protein phosphatase, EC 3.1.3.2, EC 3.1.3.48, HAAP, LMW-PTP, LMW-PTPase, Low molecular weight cytosolic acid phosphatase, low molecular weight phosphotyrosine protein phosphatase, MGC111030, MGC3499, protein tyrosine phosphatase, Red cell acid phosphatase 1 | |
| The human protein, LMW-PTP/ACP1 (GB Accession #M83653) was subcloned into pRSET to express a His6 N terminal fusion protein. This protein was expressed in E. coli and purified to homogeneity on Ni affinity matrix. | |
| 25 μg | |
| Protein Phosphatase | |
| 52 | |
| Store at -20°C. Avoid freeze/thaw cycles. | |
| IgG |
| Immunoprecipitation, Western Blot | |
| Unconjugated | |
| Phosphate buffered saline | |
| Sheep | |
| Ammonium sulfate precipitation | |
| RUO | |
| Primary | |
| Bovine, Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction