missing translation for 'onlineSavingsMsg'
Learn More

LMP2/PSMB9 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 2 publications

Brand:  Novus Biologicals NB300-625-0.025mg

 View more versions of this product

Product Code. 18500774

  • 213.00€ / 25µg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

LMP2/PSMB9 Polyclonal specifically detects LMP2/PSMB9 in Human, Mouse samples. It is validated for Western Blot.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

LMP2/PSMB9
Polyclonal
Unconjugated
PBS, 1 mg/ml BSA with 0.05% Sodium Azide
PSMB9
Mixture of synthetic peptides corresponding to residues C R(207) V I L G N/D E L P K F Y D E(220) of human and mouse proteasome 20S LMP2.
0.025 mg
Primary
Detects proteasome 20S LMP2.
Store at -20°C. Avoid freeze/thaw cycles.
IgG
Western Blot
1 mg/mL
Western Blot 1 μg/mL
beta1i, LMP2MGC70470, Low molecular mass protein 2, Macropain chain 7, Multicatalytic endopeptidase complex chain 7, proteasome (prosome, macropain) subunit, beta type, 9 (large multifunctionalpeptidase 2), proteasome catalytic subunit 1i, Proteasome chain 7, proteasome subunit beta 6i, proteasome subunit beta type-9, Proteasome subunit beta-1i, proteasome-related gene 2, PSMB6iproteasome (prosome, macropain) subunit, beta type, 9 (large multifunctionalprotease 2), Really interesting new gene 12 protein, RING12EC 3.4.25.1
Rabbit
Immunogen affinity purified
RUO
5698
Human
Purified
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only