missing translation for 'onlineSavingsMsg'
Learn More

LMP2/PSMB9 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 2 publications

213.00€

Specifications

Antigen LMP2/PSMB9
Concentration 1 mg/mL
Dilution Western Blot 1 μg/mL
Applications Western Blot
Classification Polyclonal
View More Specs

Products 1
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18500774
View Documents
Novus Biologicals
NB300-625-0.025mg
0.025 mg
213.00€
25µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

LMP2/PSMB9 Polyclonal specifically detects LMP2/PSMB9 in Human, Mouse samples. It is validated for Western Blot.
Specifications

Specifications

LMP2/PSMB9
Western Blot 1 μg/mL
Polyclonal
Purified
RUO
PBS, 1 mg/ml BSA with 0.05% Sodium Azide
5698
Mixture of synthetic peptides corresponding to residues C R(207) V I L G N/D E L P K F Y D E(220) of human and mouse proteasome 20S LMP2.
Primary
Store at -20°C. Avoid freeze/thaw cycles.
1 mg/mL
Western Blot
Unconjugated
Rabbit
Human
beta1i, LMP2MGC70470, Low molecular mass protein 2, Macropain chain 7, Multicatalytic endopeptidase complex chain 7, proteasome (prosome, macropain) subunit, beta type, 9 (large multifunctionalpeptidase 2), proteasome catalytic subunit 1i, Proteasome chain 7, proteasome subunit beta 6i, proteasome subunit beta type-9, Proteasome subunit beta-1i, proteasome-related gene 2, PSMB6iproteasome (prosome, macropain) subunit, beta type, 9 (large multifunctionalprotease 2), Really interesting new gene 12 protein, RING12EC 3.4.25.1
PSMB9
IgG
Immunogen affinity purified
Detects proteasome 20S LMP2.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.