missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ LKB1/STK11 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
213.00€ - 702.00€
Specifications
| Antigen | LKB1/STK11 |
|---|---|
| Dilution | Western Blot : 5-10 μg/mL, ELISA |
| Applications | ELISA, Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30067160
|
Novus Biologicals™
NBP3-30873-100ug |
100 μg |
702.00€
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30067099
|
Novus Biologicals™
NBP3-30873-25ug |
25 μg |
213.00€
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
LKB1/STK11 Polyclonal antibody specifically detects LKB1/STK11 in Human samples. It is validated for ELISA,Western BlotSpecifications
| LKB1/STK11 | |
| ELISA, Western Blot | |
| Unconjugated | |
| Rabbit | |
| Hypoxia Signaling, mTOR Pathway, p53 Pathway, Phospho Specific, Protein Kinase | |
| Phosphate buffered saline | |
| 6794 | |
| IgG | |
| Ammonium sulfate precipitation |
| Western Blot : 5-10 μg/mL, ELISA | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human | |
| EC 2.7.11.1, LKB1 serine/threonine kinase 11 (Peutz-Jeghers syndrome), PJS, PJS polarization-related protein LKB1, Renal carcinoma antigen NY-REN-19, serine/threonine kinase 11, serine/threonine-protein kinase 11, Serine/threonine-protein kinase LKB1, STK11 | |
| Synthetic peptide derived from the human LKB1/STK11 protein. | |
| Primary | |
| Store at -20°C. Avoid freeze/thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title