Learn More
Novus Biologicals™ LKB1/STK11 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals™ NBP3-30873-100ug
This item is not returnable.
View return policy
Description
LKB1/STK11 Polyclonal antibody specifically detects LKB1/STK11 in Human samples. It is validated for ELISA,Western Blot
Specifications
| LKB1/STK11 | |
| Polyclonal | |
| Western Blot : 5-10 μg/mL, ELISA | |
| EC 2.7.11.1, LKB1 serine/threonine kinase 11 (Peutz-Jeghers syndrome), PJS, PJS polarization-related protein LKB1, Renal carcinoma antigen NY-REN-19, serine/threonine kinase 11, serine/threonine-protein kinase 11, Serine/threonine-protein kinase LKB1, STK11 | |
| Synthetic peptide derived from the human LKB1/STK11 protein. | |
| 100 μg | |
| Hypoxia Signaling, mTOR Pathway, p53 Pathway, Phospho Specific, Protein Kinase | |
| 6794 | |
| Store at -20°C. Avoid freeze/thaw cycles. | |
| IgG |
| ELISA, Western Blot | |
| Unconjugated | |
| Phosphate buffered saline | |
| Rabbit | |
| Ammonium sulfate precipitation | |
| RUO | |
| Primary | |
| Human | |
| Purified |