missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lipid A LPS Antibody - BSA Free, Novus Biologicals™
Goat Polyclonal Antibody
210.00€ - 511.35€
Specifications
| Antigen | Lipid A LPS |
|---|---|
| Concentration | 4 mg/mL |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Form | Purified |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18041756
|
Novus Biologicals
NB100-64484-0.1ML |
0.1 mL |
222.00€ 210.00€ / 0.10mL Save 12.00€ 5% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18756813
|
Novus Biologicals
NB100-64484 |
541.00€ 511.35€ / 0.50mL Save 29.65€ 5% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||||
Description
Lipid A LPS Polyclonal specifically detects Lipid A LPS in Bacteria samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.Specifications
| Lipid A LPS | |
| Polyclonal | |
| Purified | |
| RUO | |
| Phosphate buffered saline with 0.1% Sodium Azide | |
| IgG | |
| IgG purified | |
| NB100-64484 recognizes the Lipid A or endotoxin region of LPS (Lipopolysaccharide) from E. coli. Lipid A is a glucosamine-based phospholipid which forms part of the bacterial outer membrane and anchors LPS to the surface of the bacteria. This antibody cross reacts with P. aeruginosa, K. pneumoniae, S. enteriditis, E. aerogenes, E. hermanii, Y. enterocolitica and S. sonnei. |
| 4 mg/mL | |
| Unconjugated | |
| Goat | |
| Bacteria | |
| Whole cell preparation of lipid A from E. coli O157. | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.