Learn More
Lipid A LPS Antibody - BSA Free, Novus Biologicals™
Goat Polyclonal Antibody
Brand: Novus Biologicals NB100-64484-0.1ML
This item is not returnable.
View return policy
Description
Lipid A LPS Polyclonal specifically detects Lipid A LPS in Bacteria samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.
Specifications
| Lipid A LPS | |
| Polyclonal | |
| Unconjugated | |
| Phosphate buffered saline with 0.1% Sodium Azide | |
| Whole cell preparation of lipid A from E. coli O157. | |
| 0.1 mL | |
| Primary | |
| Bacteria | |
| Purified |
| Immunocytochemistry, Immunofluorescence | |
| 4 mg/mL | |
| Immunocytochemistry/Immunofluorescence 1:10-1:2000 | |
| Goat | |
| IgG purified | |
| RUO | |
| NB100-64484 recognizes the Lipid A or endotoxin region of LPS (Lipopolysaccharide) from E. coli. Lipid A is a glucosamine-based phospholipid which forms part of the bacterial outer membrane and anchors LPS to the surface of the bacteria. This antibody cross reacts with P. aeruginosa, K. pneumoniae, S. enteriditis, E. aerogenes, E. hermanii, Y. enterocolitica and S. sonnei. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG |