missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LIF Antibody, Novus Biologicals™
Chicken Polyclonal Antibody
287.00€ - 840.00€
Specifications
| Antigen | LIF |
|---|---|
| Applications | Western Blot, ELISA |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Form | Purified |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18682179
|
Novus Biologicals
NBP3-11328-0.025mg |
0.025 mg |
287.00€
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18657046
|
Novus Biologicals
NBP3-11328 |
0.1 mg |
840.00€
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
LIF Polyclonal antibody specifically detects LIF in Rabbit samples. It is validated for Western Blot, ELISASpecifications
| LIF | |
| Polyclonal | |
| Purified | |
| RUO | |
| Rabbit | |
| CDF, D factor, DIA, differentiation inhibitory activity, differentiation stimulating factor, Differentiation-stimulating factor, Emfilermin, HILDAcholinergic differentiation factor, leukemia inhibitory factor, leukemia inhibitory factor (cholinergic differentiation factor), Melanoma-derived LPL inhibitor, MLPLI | |
| Recombinant rabbit Leukemia Inhibitory Factor | |
| Primary | |
| Store at 4°C. Do not freeze. |
| Western Blot, ELISA | |
| Unconjugated | |
| Chicken | |
| Biologically Active Proteins, Cell Cycle and Replication, Stem Cell Lines | |
| PBS | |
| 3976 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title