missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LIF Antibody, Novus Biologicals™
Chicken Polyclonal Antibody
Brand: Novus Biologicals NBP3-11328
This item is not returnable.
View return policy
Description
LIF Polyclonal antibody specifically detects LIF in Rabbit samples. It is validated for Western Blot, ELISA
Specifications
| LIF | |
| Polyclonal | |
| PBS | |
| Chicken | |
| Affinity purified | |
| RUO | |
| Primary | |
| Rabbit | |
| Purified |
| Western Blot, ELISA | |
| Unconjugated | |
| CDF, D factor, DIA, differentiation inhibitory activity, differentiation stimulating factor, Differentiation-stimulating factor, Emfilermin, HILDAcholinergic differentiation factor, leukemia inhibitory factor, leukemia inhibitory factor (cholinergic differentiation factor), Melanoma-derived LPL inhibitor, MLPLI | |
| Recombinant rabbit Leukemia Inhibitory Factor | |
| 0.1 mg | |
| Biologically Active Proteins, Cell Cycle and Replication, Stem Cell Lines | |
| 3976 | |
| Store at 4°C. Do not freeze. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction