missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LIF Antibody, Novus Biologicals™
Chicken Polyclonal Antibody
Brand: Novus Biologicals NBP3-11328-0.025mg
246.50 EUR valid until 2025-12-16
BEST PRICE on Promo! Use promo code "24090" to get your promotional price.
This item is not returnable.
View return policy
Description
LIF Polyclonal antibody specifically detects LIF in Rabbit samples. It is validated for Western Blot, ELISA
Specifications
| LIF | |
| Polyclonal | |
| PBS | |
| Chicken | |
| Affinity purified | |
| RUO | |
| Primary | |
| Rabbit | |
| Purified |
| Western Blot, ELISA | |
| Unconjugated | |
| CDF, D factor, DIA, differentiation inhibitory activity, differentiation stimulating factor, Differentiation-stimulating factor, Emfilermin, HILDAcholinergic differentiation factor, leukemia inhibitory factor, leukemia inhibitory factor (cholinergic differentiation factor), Melanoma-derived LPL inhibitor, MLPLI | |
| Recombinant rabbit Leukemia Inhibitory Factor | |
| 0.025 mg | |
| Biologically Active Proteins, Cell Cycle and Replication, Stem Cell Lines | |
| 3976 | |
| Store at 4°C. Do not freeze. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction