missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lewis Y Antibody (H18A) - Azide and BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
250.00€ - 680.00€
Specifications
| Antigen | Lewis Y |
|---|---|
| Clone | H18A |
| Concentration | 1 mg/mL |
| Applications | ELISA, Immunohistochemistry, Immunofluorescence |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18633529
|
Novus Biologicals
NBP3-09007-0.025mg |
0.025 mg |
250.00€
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18620047
|
Novus Biologicals
NBP3-09007-0.2mg |
0.2 mg |
680.00€
0.20mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Lewis Y Monoclonal antibody specifically detects Lewis Y in Human samples. It is validated for ELISA, Immunohistochemistry, Immunocytochemistry/ ImmunofluorescenceSpecifications
| Lewis Y | |
| 1 mg/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| IPCA-2, IPCA6, IPCA-6, IPCA-8, PCANAP2, PCANAP6, PCANAP8, Prostate cancer-associated protein 6, PRST, Solute carrier family 45 member 3 | |
| IgG1 κ | |
| Protein A purified |
| H18A | |
| ELISA, Immunohistochemistry, Immunofluorescence | |
| Unconjugated | |
| Mouse | |
| Cancer, Tumor Biomarkers | |
| PBS | |
| Lewis Y. | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title