Learn More
Lewis Y Antibody (H18A) - Azide and BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP3-09007-0.2mg
 View more versions of this product
Product Code. 18620047
  • 680.00€ / 0.20mg
Description
Lewis Y Monoclonal antibody specifically detects Lewis Y in Human samples. It is validated for ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Lewis Y
Monoclonal
1 mg/mL
PBS
Mouse
Protein A purified
RUO
Primary
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
IgG1 κ
ELISA, Immunohistochemistry, Immunofluorescence
H18A
Unconjugated
IPCA-2, IPCA6, IPCA-6, IPCA-8, PCANAP2, PCANAP6, PCANAP8, Prostate cancer-associated protein 6, PRST, Solute carrier family 45 member 3
Lewis Y.
0.2 mg
Cancer, Tumor Biomarkers