missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LCMT2 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
168.00€
Specifications
| Antigen | LCMT2 |
|---|---|
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1 μg/mL, ELISA 1:100-1:2000, Immunohistochemistry 5 μg/mL, Immunocytochemistry/Immunofluorescence 20 μg/mL, Immunohistochemistry-Paraffin 5 μg/mL |
| Applications | Western Blot |
| Classification | Polyclonal |
Description
LCMT2 Polyclonal specifically detects LCMT2 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
| LCMT2 | |
| Western Blot 1 μg/mL, ELISA 1:100-1:2000, Immunohistochemistry 5 μg/mL, Immunocytochemistry/Immunofluorescence 20 μg/mL, Immunohistochemistry-Paraffin 5 μg/mL | |
| Polyclonal | |
| Rabbit | |
| Human | |
| NP_055608 | |
| 9836 | |
| Antibody was raised against a 16 amino acid synthetic peptide near the amino terminus of human TYW4. The immunogen is located within the first 50 amino acids of TYW4. Amino Acid Squence: VQNTNDSSALSKRSLA | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. |
| 1 mg/mL | |
| Western Blot | |
| Unconjugated | |
| RUO | |
| PBS with 0.02% Sodium Azide | |
| EC 2.1.1.-, FLJ51321, KIAA0547tRNA-Wybutosine-synthesizing protein 4, leucine carboxyl methyltransferase 2, MGC9534, p21WAF1/CIP1 promoter-interacting protein, PPM2, tRNA wybutosine-synthesizing protein 4 homolog, tRNA yW-synthesizing protein 4 homolog, TYW4tRNA-yW synthesizing protein 4 | |
| LCMT2 | |
| IgG | |
| Affinity Purified | |
| TYW4 antibody is predicted to not cross-react with other TYW protein family members. At least two isoforms of TYW4 are known to exist; this antibody will detect only the long isoform. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title