missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LCMT2 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-76585-0.025mg
This item is not returnable.
View return policy
Description
LCMT2 Polyclonal specifically detects LCMT2 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| LCMT2 | |
| Polyclonal | |
| Unconjugated | |
| PBS with 0.02% Sodium Azide | |
| EC 2.1.1.-, FLJ51321, KIAA0547tRNA-Wybutosine-synthesizing protein 4, leucine carboxyl methyltransferase 2, MGC9534, p21WAF1/CIP1 promoter-interacting protein, PPM2, tRNA wybutosine-synthesizing protein 4 homolog, tRNA yW-synthesizing protein 4 homolog, TYW4tRNA-yW synthesizing protein 4 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 9836 | |
| Human | |
| IgG |
| Western Blot | |
| 1 mg/mL | |
| Western Blot 1 μg/mL, ELISA 1:100-1:2000, Immunohistochemistry 5 μg/mL, Immunocytochemistry/Immunofluorescence 20 μg/mL, Immunohistochemistry-Paraffin 5 μg/mL | |
| NP_055608 | |
| LCMT2 | |
| Antibody was raised against a 16 amino acid synthetic peptide near the amino terminus of human TYW4. The immunogen is located within the first 50 amino acids of TYW4. Amino Acid Squence: VQNTNDSSALSKRSLA | |
| 0.025 mg | |
| Primary | |
| TYW4 antibody is predicted to not cross-react with other TYW protein family members. At least two isoforms of TYW4 are known to exist; this antibody will detect only the long isoform. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction