missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LAMP-2/CD107b Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 7 publications
226.00€
Specifications
| Antigen | LAMP-2/CD107b |
|---|---|
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:100 - 1:2000, Immunohistochemistry 1:10 - 1:500, Immunocytochemistry/Immunofluorescence 1:10 - 1:500, Immunohistochemistry-Paraffin 1:10 - 1:500, Immunohistochemistry-Frozen |
| Applications | Western Blot |
| Classification | Polyclonal |
Description
LAMP-2/CD107b Polyclonal specifically detects LAMP-2/CD107b in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.Specifications
| LAMP-2/CD107b | |
| Western Blot 1:100 - 1:2000, Immunohistochemistry 1:10 - 1:500, Immunocytochemistry/Immunofluorescence 1:10 - 1:500, Immunohistochemistry-Paraffin 1:10 - 1:500, Immunohistochemistry-Frozen | |
| Polyclonal | |
| Rabbit | |
| Cardiovascular Biology, Cellular Markers, Chaperone Mediated Autophagy (CMA), Cognition and Behavior, Growth and Development, Immunology, Innate Immunity, Lipid and Metabolism, Lysosome Markers, Mast Cell Markers, Membrane Trafficking and Chaperones | |
| PBS with 1 mg/ml BSA with 0.05% Sodium Azide | |
| 3920 | |
| LAMP-2/CD107b Antibody was made using a synthetic Peptide: CG (400) LKRHHTGYEQF(411) | |
| Primary | |
| Store at -20°C. Avoid freeze/thaw cycles. |
| 1 mg/mL | |
| Western Blot | |
| Unconjugated | |
| RUO | |
| Human | |
| CD107 antigen-like family member B, CD107b, CD107b antigen, LAMP-2, LAMPB, LGP110, LGP-96, Lysosomal Associated Membrane Protein 2, lysosomal-associated membrane protein 2, lysosome-associated membrane glycoprotein 2, Lysosome-associated membrane protein | |
| LAMP2 | |
| IgG | |
| Affinity Purified | |
| LAMP 2 |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title