missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LAMP-2/CD107b Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 7 publications
Brand: Novus Biologicals NB300-591-0.025ml
This item is not returnable.
View return policy
Description
LAMP-2/CD107b Polyclonal specifically detects LAMP-2/CD107b in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.
Specifications
| LAMP-2/CD107b | |
| Polyclonal | |
| Unconjugated | |
| PBS with 1 mg/ml BSA with 0.05% Sodium Azide | |
| LAMP2 | |
| LAMP-2/CD107b Antibody was made using a synthetic Peptide: CG (400) LKRHHTGYEQF(411) | |
| 0.025 mL | |
| Cardiovascular Biology, Cellular Markers, Chaperone Mediated Autophagy (CMA), Cognition and Behavior, Growth and Development, Immunology, Innate Immunity, Lipid and Metabolism, Lysosome Markers, Mast Cell Markers, Membrane Trafficking and Chaperones | |
| 3920 | |
| Human | |
| IgG |
| Western Blot | |
| 1 mg/mL | |
| Western Blot 1:100 - 1:2000, Immunohistochemistry 1:10 - 1:500, Immunocytochemistry/Immunofluorescence 1:10 - 1:500, Immunohistochemistry-Paraffin 1:10 - 1:500, Immunohistochemistry-Frozen | |
| CD107 antigen-like family member B, CD107b, CD107b antigen, LAMP-2, LAMPB, LGP110, LGP-96, Lysosomal Associated Membrane Protein 2, lysosomal-associated membrane protein 2, lysosome-associated membrane glycoprotein 2, Lysosome-associated membrane protein | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| LAMP 2 | |
| Store at -20°C. Avoid freeze/thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction