missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lactalbumin Alpha Antibody (F20.16), Novus Biologicals™
Mouse Monoclonal Antibody has been used in 1 publication
Brand: Novus Biologicals NB100-2783-0.025mg
This item is not returnable.
View return policy
Description
Lactalbumin Alpha Monoclonal specifically detects Lactalbumin Alpha in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.
Specifications
| Lactalbumin Alpha | |
| Monoclonal | |
| 1.4 mg/mL | |
| Western Blot 1:100-1:2000, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1:10-1:2000 | |
| P00709 | |
| LALBA | |
| Purified casein and Lactalbumin Alpha | |
| Thiophilic Chromatography | |
| RUO | |
| Primary | |
| alpha Lactalbumin (F20.16) | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. | |
| IgM |
| Western Blot | |
| F20.16 | |
| Unconjugated | |
| PBS with 0.02% Sodium Azide | |
| alpha-lactalbumin, lactalbumin, alpha-, Lactose synthase B protein, Lysozyme-like protein 7, LYZL7, MGC138521, MGC138523 | |
| Mouse | |
| 14 kDa | |
| 0.025 mg | |
| Lipid and Metabolism | |
| 3906 | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction