missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lactalbumin Alpha Antibody (F20.16), Novus Biologicals™
Mouse Monoclonal Antibody has been used in 1 publication
178.00€
Specifications
| Antigen | Lactalbumin Alpha |
|---|---|
| Clone | F20.16 |
| Concentration | 1.4 mg/mL |
| Dilution | Western Blot 1:100-1:2000, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1:10-1:2000 |
| Applications | Western Blot |
Description
Lactalbumin Alpha Monoclonal specifically detects Lactalbumin Alpha in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.Specifications
| Lactalbumin Alpha | |
| 1.4 mg/mL | |
| Western Blot | |
| Unconjugated | |
| Mouse | |
| Lipid and Metabolism | |
| PBS with 0.02% Sodium Azide | |
| alpha-lactalbumin, lactalbumin, alpha-, Lactose synthase B protein, Lysozyme-like protein 7, LYZL7, MGC138521, MGC138523 | |
| LALBA | |
| IgM | |
| Thiophilic Chromatography | |
| alpha Lactalbumin (F20.16) |
| F20.16 | |
| Western Blot 1:100-1:2000, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1:10-1:2000 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| P00709 | |
| 3906 | |
| Purified casein and Lactalbumin Alpha | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. | |
| 14 kDa |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title