Learn More
Epredia™ Lab Vision™ Progesterone Receptor Rabbit Monoclonal Antibody
Progesterone Receptor, Rabbit Monoclonal Antibody is part of a comprehensive antibody portfolio. Studies show rabbit monoclonal antibodies are more sensitive and specific than other available markers.
322.00€ - 1466.35€
Specifications
| Antigen | Progesterone Receptor (SP2) |
|---|---|
| Clone | SP2 |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
| Product Code | Brand | Quantity | Form | Price | Quantity & Availability | ||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Form | Price | Quantity & Availability | ||||
|
12623677
|
Epredia™
RM-9102-S0 |
100 μL | Concentrated | ||||||
|
12633677
|
Epredia™
RM-9102-S1 |
500 μL | Concentrated |
1165.00€ 1059.96€ / 500µL Save 105.04€ 9% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
|
12613677
|
Epredia™
RM-9102-S |
1 mL | Concentrated |
1715.00€ 1466.35€ / 1mL Save 248.65€ 14% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
|
12683667
|
Epredia™
RM-9102-R7 |
7 mL | Prediluted |
692.00€ 608.81€ / 7mL Save 83.19€ 12% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
|
12603677
|
Epredia™
RM-9102-RQ |
12 mL | Ready-to-Use |
947.00€ 800.96€ / 12mL Save 146.04€ 15% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
The progesterone receptor (PgR) is an estrogen-regulated protein. It has been proposed that expression of PgR determination indicates a responsive estrogen receptor (ER) pathway, and therefore, may predict likely response to endocrine therapy in human breast cancer. A number of studies have shown that PgR determination provides supplementary information to ER, both in predicting response to endocrine therapy and estimating survival. PgR has proved superior to ER as a prognostic indicator in some studies.Host Species: Rabbit
Clone: SP2
Isotype: IgG
Species Reactivity: Human. Others not tested.
Epitope: 412-526aa
Immunogen: Recombinant protein encoding human progesterone receptor 412-526aa
Molecular Weight: 116 + 81kDa
Positive Control: Breast CA, T47D cells
Cellular Localization: Nuclear
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
- Western Blotting
Specifications
| Progesterone Receptor (SP2) | |
| Monoclonal | |
| Rabbit | |
| Cancer and Tumor Biology | |
| Primary |
| SP2 | |
| Unconjugated | |
| IVD | |
| Recombinant protein encoding human progesterone receptor 412-526 aa |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.