missing translation for 'onlineSavingsMsg'
Learn More

Epredia™ Lab Vision™ Progesterone Receptor Rabbit Monoclonal Antibody

Progesterone Receptor, Rabbit Monoclonal Antibody is part of a comprehensive antibody portfolio. Studies show rabbit monoclonal antibodies are more sensitive and specific than other available markers.

Brand:  Epredia™ RM-9102-S

 View more versions of this product

Product Code. 12613677

  • ONLINE EXCLUSIVE PRICE
    1715.00€
    1466.35€ / 1mL
    Save 248.65€
    14% Off

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

The progesterone receptor (PgR) is an estrogen-regulated protein. It has been proposed that expression of PgR determination indicates a responsive estrogen receptor (ER) pathway, and therefore, may predict likely response to endocrine therapy in human breast cancer. A number of studies have shown that PgR determination provides supplementary information to ER, both in predicting response to endocrine therapy and estimating survival. PgR has proved superior to ER as a prognostic indicator in some studies.

Host Species: Rabbit

Clone: SP2

Isotype: IgG

Species Reactivity: Human. Others not tested.

Epitope: 412-526aa

Immunogen: Recombinant protein encoding human progesterone receptor 412-526aa

Molecular Weight: 116 + 81kDa

Positive Control: Breast CA, T47D cells

Cellular Localization: Nuclear

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Progesterone Receptor (SP2)
Monoclonal
Unconjugated
Recombinant protein encoding human progesterone receptor 412-526 aa
IVD
Primary
Concentrated
Western Blot
SP2
Rabbit
1 mL
Cancer and Tumor Biology
Human
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.