missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KSV Cyclin D Antibody (94B) - BSA Free, Novus Biologicals™
Rat Monoclonal Antibody
190.00€ - 453.00€
Specifications
| Antigen | KSV Cyclin D |
|---|---|
| Clone | 94B |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:100 - 1:2000, Immunohistochemistry 1:10 - 1:500, Immunocytochemistry/ Immunofluorescence 1:10 - 1:500, Immunoprecipitation 1:10 - 1:500 |
| Applications | Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18477439
|
Novus Biologicals
NB100-2621-0.025mg |
0.025 mg |
190.00€
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18470379
|
Novus Biologicals
NB100-2621 |
0.1 mg |
453.00€
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
KSV Cyclin D Monoclonal antibody specifically detects KSV Cyclin D in Virus samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, ImmunoprecipitationSpecifications
| KSV Cyclin D | |
| 1 mg/mL | |
| Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation | |
| Unconjugated | |
| Rat | |
| Cancer, Cell Cycle and Replication, Core ESC Like Genes, mTOR Pathway, Stem Cell Markers, Wnt Signaling Pathway | |
| PBS | |
| Recombinant fusion protein produced from a clone from KSV. | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| 94B | |
| Western Blot 1:100 - 1:2000, Immunohistochemistry 1:10 - 1:500, Immunocytochemistry/ Immunofluorescence 1:10 - 1:500, Immunoprecipitation 1:10 - 1:500 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Virus | |
| Kaposi's Sarcoma Virus Type D Cyclin | |
| IgG | |
| Protein G purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title