missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KSV Cyclin D Antibody (94B) - BSA Free, Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals NB100-2621
This item is not returnable.
View return policy
Description
KSV Cyclin D Monoclonal antibody specifically detects KSV Cyclin D in Virus samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation
Specifications
| KSV Cyclin D | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:100 - 1:2000, Immunohistochemistry 1:10 - 1:500, Immunocytochemistry/ Immunofluorescence 1:10 - 1:500, Immunoprecipitation 1:10 - 1:500 | |
| Kaposi's Sarcoma Virus Type D Cyclin | |
| Recombinant fusion protein produced from a clone from KSV. | |
| 0.1 mg | |
| Cancer, Cell Cycle and Replication, Core ESC Like Genes, mTOR Pathway, Stem Cell Markers, Wnt Signaling Pathway | |
| Virus | |
| Purified |
| Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation | |
| 94B | |
| Unconjugated | |
| PBS | |
| Rat | |
| Protein G purified | |
| RUO | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction