missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KIR3DL1 Antibody (6D9F6) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
203.00€ - 552.00€
Specifications
| Antigen | KIR3DL1 |
|---|---|
| Clone | 6D9F6 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:100-1:2000, ELISA 1:100-1:2000, Immunohistochemistry 1:10-1:1000, Immunohistochemistry-Paraffin 1:200 - 1:1000 |
| Applications | Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18253441
|
Novus Biologicals
NBP2-61811 |
0.1 mL |
552.00€
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18609016
|
Novus Biologicals
NBP2-61811-0.025ml |
0.025 mL |
203.00€
0.03mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
KIR3DL1 Monoclonal specifically detects KIR3DL1 in Human, Rat, Monkey samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| KIR3DL1 | |
| 1 mg/mL | |
| Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Mouse | |
| Human, Rat, Monkey | |
| 3811 | |
| Purified recombinant fragment of human KIR3DL1 (AA: extra 22-340) expressed in Hek293 cells. | |
| Primary | |
| 49 kDa |
| 6D9F6 | |
| Western Blot 1:100-1:2000, ELISA 1:100-1:2000, Immunohistochemistry 1:10-1:1000, Immunohistochemistry-Paraffin 1:200 - 1:1000 | |
| Monoclonal | |
| Purified | |
| Immunology | |
| AMB11, CD158 antigen-like family member E, CD158E, CD158e antigen, CD158e1, CD158e1/2, CD158e2, cl-11, cl-2, HLA-BW4-specific inhibitory NK cell receptor, killer cell immunoglobulin-like receptor 3DL1, killer cell immunoglobulin-like receptor, three domains, long cytoplasmic tail1,nkat3, killer Ig receptor, KIR antigen 3DL1, KIR3DL1/S1, KIR3DS1, MGC119726, MGC119728, MGC126589, MGC126591, MHC class I NK cell receptor, Natural killer-associated transcript 3, NKAT-3, NKAT3KIR, NKB1B, NKB1CD158E1, NK-receptor, p70 killer cell inhibitory receptor, p70 natural killer cell receptor clones CL-2/CL-11, p70 NK receptor CL-2/CL-11 | |
| KIR3DL1 | |
| IgG2b | |
| Protein G purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title