missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KIF17 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB600-1319-0.025ml
This item is not returnable.
View return policy
Description
KIF17 Polyclonal specifically detects KIF17 in Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence, Microarray.
Specifications
| KIF17 | |
| Polyclonal | |
| Western Blot 1:1000, Immunocytochemistry/Immunofluorescence 1:100, Microarray | |
| KIAA1405KIF3XKIF17 variant protein, KIF17B, KIF3-related motor protein, kinesin family member 17, kinesin-like protein KIF17 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| KIF17 | |
| Store at -20°C. Avoid freeze/thaw cycles. |
| Western Blot | |
| Unconjugated | |
| 10mM PBS (pH 7.4) with 15mM Sodium Azide | |
| KIF17 | |
| Synthetic peptide conjugated to KLH, corresponding to amino acids 589-606 of Mouse KIF17. | |
| 0.025 mL | |
| Cytoskeleton Markers | |
| 57576 | |
| Mouse | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction