missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KIF17 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
187.00€
Specifications
| Antigen | KIF17 |
|---|---|
| Dilution | Western Blot 1:1000, Immunocytochemistry/Immunofluorescence 1:100, Microarray |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
Description
KIF17 Polyclonal specifically detects KIF17 in Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence, Microarray.Specifications
| KIF17 | |
| Western Blot | |
| Unconjugated | |
| RUO | |
| Mouse | |
| KIAA1405KIF3XKIF17 variant protein, KIF17B, KIF3-related motor protein, kinesin family member 17, kinesin-like protein KIF17 | |
| KIF17 | |
| IgG | |
| Affinity Purified | |
| KIF17 |
| Western Blot 1:1000, Immunocytochemistry/Immunofluorescence 1:100, Microarray | |
| Polyclonal | |
| Rabbit | |
| Cytoskeleton Markers | |
| 10mM PBS (pH 7.4) with 15mM Sodium Azide | |
| 57576 | |
| Synthetic peptide conjugated to KLH, corresponding to amino acids 589-606 of Mouse KIF17. | |
| Primary | |
| Store at -20°C. Avoid freeze/thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title