missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KHDRBS3 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
190.00€ - 550.00€
Specifications
| Antigen | KHDRBS3 |
|---|---|
| Dilution | Western Blot 1:500 - 1:2000, ELISA, Immunocytochemistry/Immunofluorescence 1:50 - 1:100 |
| Applications | ELISA, Western Blot, Immunocytochemistry/Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30231539
|
Novus Biologicals
NBP3-38301-20ul |
20 μL |
190.00€
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30227276
|
Novus Biologicals
NBP3-38301-100ul |
100 μL |
550.00€
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
KHDRBS3 Polyclonal antibody specifically detects KHDRBS3 in Human,Mouse samples. It is validated for ELISA,Western Blot,Immunocytochemistry/ImmunofluorescenceSpecifications
| KHDRBS3 | |
| ELISA, Western Blot, Immunocytochemistry/Immunofluorescence | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse | |
| Etle, etoile, KH domain containing, RNA binding, signal transduction associated 3, SALPRNA-binding protein T-Star, Sam68-like mammalian protein 2, Sam68-like phosphotyrosine protein, Sam68-like phosphotyrosine protein, T-STAR, SLM-2KH domain-containing, RNA-binding, signal transduction-associated protein 3, SLM2TSTAR, T-STAR | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 177-346 of human KHDRBS3 (NP_006549.1).,, Sequence:, GSENADVPVVRGKPTLRTRGVPAPAITRGRGGVTARPVGVVVPRGTPTPRGVLSTRGPVSRGRGLLTPRARGVPPTGYRPPPPPPTQETYGEYDYDDGYGTAYDEQSYDSYDNSYSTPAQSGADYYDYGHGLSEETYDSYGQEEWTNSRHKAPSARTAKGVYRDQPYGRY | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
| Western Blot 1:500 - 1:2000, ELISA, Immunocytochemistry/Immunofluorescence 1:50 - 1:100 | |
| Polyclonal | |
| Purified | |
| RUO | |
| PBS (pH 7.3), 50% glycerol | |
| 10656 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title