missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KHDRBS3 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-38301-100ul
This item is not returnable.
View return policy
Description
KHDRBS3 Polyclonal antibody specifically detects KHDRBS3 in Human,Mouse samples. It is validated for ELISA,Western Blot,Immunocytochemistry/Immunofluorescence
Specifications
| KHDRBS3 | |
| Polyclonal | |
| Western Blot 1:500 - 1:2000, ELISA, Immunocytochemistry/Immunofluorescence 1:50 - 1:100 | |
| Etle, etoile, KH domain containing, RNA binding, signal transduction associated 3, SALPRNA-binding protein T-Star, Sam68-like mammalian protein 2, Sam68-like phosphotyrosine protein, Sam68-like phosphotyrosine protein, T-STAR, SLM-2KH domain-containing, RNA-binding, signal transduction-associated protein 3, SLM2TSTAR, T-STAR | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 177-346 of human KHDRBS3 (NP_006549.1).,, Sequence:, GSENADVPVVRGKPTLRTRGVPAPAITRGRGGVTARPVGVVVPRGTPTPRGVLSTRGPVSRGRGLLTPRARGVPPTGYRPPPPPPTQETYGEYDYDDGYGTAYDEQSYDSYDNSYSTPAQSGADYYDYGHGLSEETYDSYGQEEWTNSRHKAPSARTAKGVYRDQPYGRY | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Purified |
| ELISA, Western Blot, Immunocytochemistry/Immunofluorescence | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 10656 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction