missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KAP1 Antibody (20C1), Novus Biologicals™
Mouse Monoclonal Antibody
200.00€
Specifications
| Antigen | KAP1 |
|---|---|
| Clone | 20C1 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1 μg/mL, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 2 - 4 μg/mL, Immunoprecipitation 1:10 - 1:500, Immunohistochemistry-Paraffin 1:10-1:500, Gel Super Shift Assays 1:1 - 1:100, Chromatin Immunoprecipitation (ChIP) 1:10-1:500 |
| Applications | Western Blot |
Description
KAP1 Monoclonal specifically detects KAP1 in Human, Mouse samples. It is validated for Western Blot, Chromatin Immunoprecipitation, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Gel Super Shift Assays.Specifications
| KAP1 | |
| 1 mg/mL | |
| Western Blot | |
| Unconjugated | |
| Mouse | |
| Chromatin Research, DNA Repair, DNA replication Transcription Translation and Splicing, Protein Kinase, Signal Transduction, Transcription Factors and Regulators, Zinc Finger | |
| PBS with 1mg/ml BSA with 0.05% Sodium Azide | |
| 10155 | |
| Recombinant protein corresponding to residues 60-383 of the human TIF1 beta protein. | |
| Primary | |
| Store at -20°C. Avoid freeze/thaw cycles. |
| 20C1 | |
| Western Blot 1 μg/mL, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 2 - 4 μg/mL, Immunoprecipitation 1:10 - 1:500, Immunohistochemistry-Paraffin 1:10-1:500, Gel Super Shift Assays 1:1 - 1:100, Chromatin Immunoprecipitation (ChIP) 1:10-1:500 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| E3 SUMO-protein ligase TRIM28, EC 6.3.2.-, FLJ29029, KAP-1, KAP1KRAB-associated protein 1, KRIP-1, Nuclear corepressor KAP-1, RNF96KRAB-interacting protein 1, TF1B, TIF1-beta, TIF1BRING finger protein 96, transcription intermediary factor 1-beta, transcriptional intermediary factor 1-beta, tripartite motif containing 28, tripartite motif-containing 28, Tripartite motif-containing protein 28 | |
| TRIM28 | |
| IgG1 | |
| Protein G purified | |
| TIF1 beta (20C1) |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title