missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KAP1 Antibody (20C1), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB100-74549-0.025mg
This item is not returnable.
View return policy
Description
KAP1 Monoclonal specifically detects KAP1 in Human, Mouse samples. It is validated for Western Blot, Chromatin Immunoprecipitation, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Gel Super Shift Assays.
Specifications
| KAP1 | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1 μg/mL, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 2 - 4 μg/mL, Immunoprecipitation 1:10 - 1:500, Immunohistochemistry-Paraffin 1:10-1:500, Gel Super Shift Assays 1:1 - 1:100, Chromatin Immunoprecipitation (ChIP) 1:10-1:500 | |
| E3 SUMO-protein ligase TRIM28, EC 6.3.2.-, FLJ29029, KAP-1, KAP1KRAB-associated protein 1, KRIP-1, Nuclear corepressor KAP-1, RNF96KRAB-interacting protein 1, TF1B, TIF1-beta, TIF1BRING finger protein 96, transcription intermediary factor 1-beta, transcriptional intermediary factor 1-beta, tripartite motif containing 28, tripartite motif-containing 28, Tripartite motif-containing protein 28 | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| TIF1 beta (20C1) | |
| Store at -20°C. Avoid freeze/thaw cycles. | |
| IgG1 |
| Western Blot | |
| 20C1 | |
| Unconjugated | |
| PBS with 1mg/ml BSA with 0.05% Sodium Azide | |
| TRIM28 | |
| Recombinant protein corresponding to residues 60-383 of the human TIF1 beta protein. | |
| 0.025 mg | |
| Chromatin Research, DNA Repair, DNA replication Transcription Translation and Splicing, Protein Kinase, Signal Transduction, Transcription Factors and Regulators, Zinc Finger | |
| 10155 | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction