missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Kallikrein 6/Neurosin Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
199.00€ - 450.00€
Specifications
| Antigen | Kallikrein 6/Neurosin |
|---|---|
| Dilution | Western Blot 1:1000, Immunocytochemistry/Immunofluorescence 1:20 |
| Applications | Western Blot, Immunocytochemistry, Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18293980
|
Novus Biologicals
NBP1-06497SS |
0.025 mL |
199.00€
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18201580
|
Novus Biologicals
NBP1-06497 |
0.1 mL |
450.00€
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Kallikrein 6/Neurosin Polyclonal specifically detects Kallikrein 6/Neurosin in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.Specifications
| Kallikrein 6/Neurosin | |
| Western Blot, Immunocytochemistry, Immunofluorescence | |
| Unconjugated | |
| Rabbit | |
| Cancer, Cell Biology, Cellular Markers, Cytoskeleton Markers, Endothelial Cell Markers, Extracellular Matrix, Neurodegeneration, Neuroscience, Signal Transduction | |
| Whole antisera with 0.1% Sodium Azide | |
| Bssp, EC 3.4.21, EC 3.4.21.-, hK6, kallikrein 6 (neurosin, zyme), kallikrein-6, kallikrein-related peptidase 6, Klk7, neurosin, Protease M, protease, serine, 18, PRSS18, PRSS9MGC9355, Serine protease 18, Serine protease 9, SP59, Zyme | |
| KLK6 | |
| IgG | |
| Unpurified | |
| Human. Immunogen sequence has 85% identity with canine, 82% identity with horse and 80% identity with bovine. |
| Western Blot 1:1000, Immunocytochemistry/Immunofluorescence 1:20 | |
| Polyclonal | |
| Antisera | |
| RUO | |
| Human | |
| Q92876 | |
| 5653 | |
| Full length human Kallikrein 6 protein expressed in E. coli. [Swiss-Prot# Q92876] | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| 38 kDa |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title