missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Kallikrein 6/Neurosin Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-06497SS
This item is not returnable.
View return policy
Description
Kallikrein 6/Neurosin Polyclonal specifically detects Kallikrein 6/Neurosin in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.
Specifications
| Kallikrein 6/Neurosin | |
| Polyclonal | |
| Western Blot 1:1000, Immunocytochemistry/Immunofluorescence 1:20 | |
| Q92876 | |
| KLK6 | |
| Full length human Kallikrein 6 protein expressed in E. coli. [Swiss-Prot# Q92876] | |
| Unpurified | |
| RUO | |
| Primary | |
| Human. Immunogen sequence has 85% identity with canine, 82% identity with horse and 80% identity with bovine. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunocytochemistry, Immunofluorescence | |
| Unconjugated | |
| Whole antisera with 0.1% Sodium Azide | |
| Bssp, EC 3.4.21, EC 3.4.21.-, hK6, kallikrein 6 (neurosin, zyme), kallikrein-6, kallikrein-related peptidase 6, Klk7, neurosin, Protease M, protease, serine, 18, PRSS18, PRSS9MGC9355, Serine protease 18, Serine protease 9, SP59, Zyme | |
| Rabbit | |
| 38 kDa | |
| 0.025 mL | |
| Cancer, Cell Biology, Cellular Markers, Cytoskeleton Markers, Endothelial Cell Markers, Extracellular Matrix, Neurodegeneration, Neuroscience, Signal Transduction | |
| 5653 | |
| Human | |
| Antisera |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction