missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ITK Mouse anti-Human, CoraLite 488, Clone: 1F9D8, Proteintech
Mouse Monoclonal Antibody
145.00€ - 450.00€
Specifications
| Antigen | ITK |
|---|---|
| Clone | 1F9D8 |
| Applications | Immunofluorescence |
| Classification | Monoclonal |
| Conjugate | CoraLite 488 |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
17741309
|
Proteintech
CL488-67703-20UL |
20 μL |
145.00€
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
17689876
|
Proteintech
CL488-67703-100UL |
100 μL |
450.00€
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
ITK Monoclonal antibody apecifically detetects ITK antigen in Human Samples. It is validated for Immunofluorescence.Specifications
| ITK | |
| Immunofluorescence | |
| CoraLite 488 | |
| Mouse | |
| PBS with 50% Glycerol, 0.05% Proclin300, 0.5% BSA, pH 7.3. | |
| 3702 | |
| ITK fusion protein Ag17087 | |
| Primary | |
| Store at -20°C. Avoid exposure to light. |
| 1F9D8 | |
| Monoclonal | |
| Liquid | |
| Human | |
| BC109077 | |
| ITK | |
| IgG2a | |
| Protein A purification |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title