missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ITK Mouse anti-Human, CoraLite 488, Clone: 1F9D8, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech CL488-67703-100UL
This item is not returnable.
View return policy
Description
ITK Monoclonal antibody apecifically detetects ITK antigen in Human Samples. It is validated for Immunofluorescence.Specifications
| ITK | |
| Monoclonal | |
| 1000 μg/mL | |
| PBS with 50% Glycerol, 0.05% Proclin300, 0.5% BSA, pH 7.3. | |
| ITK | |
| ITK fusion protein Ag17087 | |
| 100 μL | |
| 3702 | |
| Store at -20°C. Avoid exposure to light. | |
| IgG2a |
| Immunofluorescence | |
| 1F9D8 | |
| CoraLite 488 | |
| BC109077 | |
| Mouse | |
| Protein A purification | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction